missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talc, particle size: -200 mesh (<75 μm), Thermo Scientific Chemicals
£14.80 - £112.00
Chemical Identifiers
CAS | 14807-96-6 |
---|---|
Molecular Formula | H2Mg3O12Si4 |
Molecular Weight (g/mol) | 379.26 |
MDL Number | MFCD00792903 |
InChI Key | KNXVOGGZOFOROK-UHFFFAOYSA-N |
Synonym | magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate |
PubChem CID | 16211421 |
IUPAC Name | dioxosilane;oxomagnesium;hydrate |
SMILES | [Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10503244
|
Thermo Scientific Acros
222735000 |
500 g | Plastic bottle | ||||||
10772071
|
Thermo Scientific Acros
222730025 |
2.5 kg | Plastic bottle | ||||||
11994321
|
Thermo Scientific Acros
222730100 |
10 kg | Plastic drum | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14807-96-6 | |
379.26 | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
16211421 | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
dioxosilane;oxomagnesium;hydrate |
Specifications
800.0°C | |
White | |
H2Mg3O12Si4 | |
MFCD00792903 | |
magnesium silicate, hydrous, talc powder, talc, cp, talc,-350 mesh, talc, powder, 10 mum, 3mgo.4o2si.h2o, talc, vetec tm reagent grade, talc, tested according to ph.eur., magnesium silicate monohydrate talc, tris @magnesium oxide tetrakis silica hydrate | |
KNXVOGGZOFOROK-UHFFFAOYSA-N | |
dioxosilane;oxomagnesium;hydrate | |
16211421 | |
Typical chemical properties: SiO2: 59.3%, MgO: 31.5%, Al2O3: 1.9%, Fe2O3: 1.0%, CaO: 0.6% | |
Talc |
14807-96-6 | |
Very Fine Powder | |
Mg3[Si4O10(OH)2] | |
15,9168 | |
Solubility in water: insoluble. Other solubilities: insoluble in cold acids and alkalies | |
[Mg++].[Mg++].[Mg++].O[Si]([O-])=O.O[Si]([O-])=O.[O-][Si]([O-])=O.[O-][Si]([O-])=O | |
379.26 | |
379.28 | |
0.2% (105°C) max. |
Safety and Handling
missing translation for 'einecsNumber' : 238-877-9
RUO – Research Use Only