missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Glass Specimen Vials

For use with push-in caps. Fisherbrand™ Glass Specimen Vials are manufactured from clear Type 1B neutral glass.

£120.12 - £146.00

Specifications

Color Clear
Material Borosilicate Glass
Style Tall form
Base Style Flat
Closure Material Polyethylene
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
14863542
View Documents
Fisherbrand™
155063 + 105057
7 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15324779
View accessory products View Documents
Fisherbrand™
15324779
3.5 mL
£146.00
Pack of 666
Estimated Shipment: 27-03-2025
to see stock.
Add to basket
 
Description

Description

  • Flat bottom
  • Borosilicate glass
  • Suitable for media, diagnostic, storage, display and sample collection applications
Specifications

Specifications

Clear
Tall form
Polyethylene
Flat Bottom Vial
Borosilicate Glass
Flat
Push-in Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.