missing translation for 'onlineSavingsMsg'
Learn More

Liebherr™ Tall Heavy-Duty Lab Fridge

For secure, accurate and long-lasting fridge storage of temperature-sensitive products, the Liebherr™ Tall Heavy-Duty Lab Fridge is fully-equipped for lab applications with Liebherr build-quality you can rely on. EN 60068 temperature mapping and SmartMonitoring options.

Brand:  Liebherr™ LKPv 6523

 View more versions of this product

Product Code. 15280469

  • £6290.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • White exterior, 304-grade stainless steel interior, fan-assisted refrigeration.
  • Refrigerant R 290, top-mounted integral refrigeration
  • 125mm castors for easy cleaning. Front castors with brake.
  • Digital controller with 0,1°C resolution, password keypad lock, RS 485 interface and volt-free alarm contact
  • Controller battery-powered for 72hrs in case of power failure
  • Solid-state compressor relay for maximum reliability and temperature stability
  • Alarm log of last 3 events with min/max temperatures, duration etc,
  • Wire-on-tube condenser with removable filter for reliability and low-maintenance
  • Door lock, reversible hinges, self-closing door with 90° dwell position
  • 60 kg shelf load, access port for external temperature probe
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Liebherr™ Tall Heavy-Duty Lab Fridge > Capacity: 597 L, Glass Door

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.