missing translation for 'onlineSavingsMsg'
Learn More

Cytiva TALON™ Superflow Medium

Offers different selectivity compared to nickel-charged media. Cytiva TALON™ Superflow Medium used for purification of histidine-tagged recombinant proteins by IMAC.

Brand:  Cytiva 28-9574-99


 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11658893

This item is not returnable. View return policy

Description

Description

Features:

  • Ligand - TALON Superflow Cobalt-IMAC
  • Matrix - Cross-linked agarose, 6%
  • Particle Size - 60 -160μm
  • Binding Capacity/mL - Up to 20 mg histidine-tagged protein
  • pH stability working range - 3 to 12
  • pH stability Cleaning-in-Place (CIP) - 2 - 14 Co2+stripped medium
  • Complete packsize - 10mL

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mL
4°C to 8°C
Talon Superflow Media
6% Cross-Linked Agarose
20% Ethanol
For PurifiCatIon of Histidine-Tagged Recombinant Pby Immobilized Metal Affinity Chromatography
Up to 20 mg Histidine-tagged Protein
60 to 160 μm
3 to 12
Talon Superflow Cobalt-IMAC
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers