Learn More
Thermo Scientific™ Tango™ CXCR7-bla U2OS Cells
The Tango™ CXCR7-bla U2OS cells contain the human Chemokine (C-X-C motif) receptor 7 (CXCR7) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1832
Product Code. 10481215
  • £10340.00 / Each
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Tango™ CXCR7-bla U2OS cells contain the human Chemokine (C-X-C motif) receptor 7 (CXCR7) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element. The Tango™ CXCR7-bla U2OS cells are functionally validated for Z' and EC50 concentrations of SDF1a. In addition, Tango™ CXCR7-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NM_020311.2
Dividing Cells
Native Tail
Fluorescence
CXCR7
Tango™ CXCR7-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ CXCR7-bla U2OS cells, Tango™ CXCR7-bla U2OS assay protocol, certificate of analysis
GeneBLAzer™, Tango™
End Point
CXCR7
U2OS
GPCR
Cell-based beta-arrestin recruitment
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ CXCR7-bla U2OS cells
• Tango™ CXCR7-bla U2OS Assay Protocol
• Certificate of Analysis
The Tango™ CXCR7-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Tango™ CXCR7-bla U2OS Cells, 8 x 106 Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
FRET
G-Protein Coupled Receptor (GPCR)
Single Cell Clone Origin
8 x 106 Cells
U2OS
U2OS Cell
Tango™