Learn More
Invitrogen™ Tango™ F2RL1-bla U2OS Cells
Tango™ F2RL1-bla U2OS Cells
Brand: Invitrogen™ K1830
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The Tango™ F2RL1-bla U2OS cells contain the human Coagulation factor II (thrombin) receptor-like 1 (F2RL1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin⁄TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.The Tango™ F2RL1-bla U2OS cells are functionally validated for Z' and EC50 concentrations of SLIGRL. In addition, Tango™ F2RL1-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
Specifications
Liquid Nitrogen | |
Fluorescent | |
Tango™ F2RL1-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ F2RL1-bla U2OS cells, Tango™ F2RL1-bla U2OS assay protocol, certificate of analysis | |
U2OS |
Tango™ F2RL1-bla U2OS Cells, 8 x 106 Cells, ∽8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage | |
Pharma and biopharma, drug discovery and development, target and lead identification and validation, G-protein coupled receptor biology, cell-based GPCR reporter assays | |
8 x 106 Cells | |
U2OS Cell |
For Research Use Only. Not for use in diagnostic procedures.