Learn More
Thermo Scientific™ Tango™ H4-bla U2OS
The Tango™ H4-bla U2OS cells contain the human Histamine Receptor 4 (H4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand:  Thermo Scientific™ K1465
Product Code. 10185273
  • £10780.00 / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
The Tango™ H4-bla U2OS cells contain the human Histamine Receptor 4 (H4) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ H4-bla U2OS cells are functionally validated for Z' and EC50 concentrations of 4-Methylhistamine. In addition, Tango™ H4-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary.
For Academic pricing please login or contact us at discoverysciences@invitrogen.com.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NP_067637.2
Division-Arrested Cells
Tail from AVPR2 GPCR
Fluorescence
HRH4
Single Cell Clone Origin
8 x 108 Cells
BLA (Beta-Lactamase)
HRH4, H4
U2OS
GPCR
Cell-based beta-arrestin recruitment
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol.
Includes:
• Tango™ H4-bla U2OS cells
• Tango™ H4-bla U2OS Assay Protocol
• Certificate of Analysis
The Tango™ H4-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt.
Tango™ H4-bla U2OS, 8 x 106 Cells, ∽8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, U2OS Cell Line, Fluorescent Detection Method, GPCRs Druggable Target, BLA (Beta-Lactamase) Reporter, Liquid Nitrogen Storage
FRET
Tango™ H4-bla U2OS cells (LiveBLAzer™-FRET B/G loading kit, solution D and other materials are required separately; please refer to the protocol). Includes: Tango™ H4-bla U2OS cells, Tango™ H4-bla U2OS assay protocol, certificate of analysis
GeneBLAzer™, Tango™
End Point
U2OS
U2OS Cell
Tango™