Learn More
Thermo Scientific™ Tango™ M1-bla U2OS Cells
Tango™ M1-bla U2OS cells contain the human muscarinic M1 receptor (CHRM1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™ K1861
Description
Tango™ M1-bla U2OS cells contain the human muscarinic M1 receptor (CHRM1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase reporter gene under the control of a UAS response element. The Tango™ M1-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of oxotremorine M. In addition, Tango™ M1-bla U2OS cells have been tested for assay performance under variable conditions.
Specifications
NM_000738 | |
Dividing Cells | |
Native Tail | |
FRET | |
Single Cell Clone Origin | |
8 x 106 Cells | |
BLA (Beta-Lactamase) | |
Dividing Cells | |
Tango™ |
Cell-based beta-arrestin recruitment | |
Tango™ cell lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. | |
Fluorescence | |
G-Protein Coupled Receptor (GPCR) | |
Tango™ | |
End Point | |
M1 | |
U2OS | |
GPCR |
For Research Use Only. Not for use in diagnostic procedures.