Learn More
Corning™ Tank Liners
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
Brand:  Corning™ 91-300-35
 View more versions of this product
Product Code. 17213594
  • £72.30 / Each
Explore more special offers
Description
Description
  • Made from low-density polyethylene (LDPE)
  • Eliminate costs associated with cleaning and validation of tanks
  • Decrease labor costs and increase turnaround time
  • Eliminate risks associated with cross contamination
  • Help extend life of reuseable tanks, resulting in reduced capital expenditures
  • Gamma irradiated, individually packaged
  • Choice of gusseted and non-gusseted available
  • Available in many sizes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 L
0.0528 gal.
91.44 cm
55.88 cm
22 in. x 36 in. (55.9 cm x 91.4 cm)
LDPE
36 in.
22 in.
Cylindrical
Gusseted Tank Liner