missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum(V) bromide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
50 g
250 g
Description
The Tantalum Bromide Cluster is used for the preparation of heavy-atom derivatives for structure determination of biological macromolecules by X-ray analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 348.8°C |
Melting Point | 265°C |
Color | Yellow |
Physical Form | Powder |
Quantity | 2 g |
UN Number | UN3260 |
Formula Weight | 580.49 |
Percent Purity | 99.9% |
Odor | Pungent |
Sensitivity | Moisture Sensitive |
Afficher plus |
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu