Learn More
Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon

Specifications
Specifications
Boiling Point | 242°C |
Melting Point | 216°C |
Color | White to Yellow |
Physical Form | Powder |
Quantity | 5 g |
UN Number | UN3260 |
Merck Index | 14,9054 |
Formula Weight | 358.21 |
Percent Purity | 99.995% |
Grade | Puratronic™ |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z30K013 | Certificate of Analysis | 31/12/2024 | 043731.06,043731.14,043731.22 |
W06K047 | Certificate of Analysis | 07/10/2024 | 043731.06,043731.14,043731.22 |
M02K040 | Certificate of Analysis | 10/01/2024 | 043731.06,043731.14,043731.22 |
T02G022 | Certificate of Analysis | 22/11/2022 | 043731.06,043731.14,043731.22 |
Z27C088 | Certificate of Analysis | 18/11/2021 | 043731.06,043731.14,043731.22 |
Safety and Handling


- Tantalum(V) chloride, anhydrous, Puratronic
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.