missing translation for 'onlineSavingsMsg'
Learn More
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11317057

Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11317057
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11317057

Brand: Thermo Scientific Alfa Aesar 010880.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tantalum(V) chloride is used to form mixed chloro-aryloxide compounds, such as [Ta(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry. It is also used as a starting material for new edge-bridged octahedral M6 cluster compounds such as CsErTa6Cl18.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]

Specifications

Boiling Point 242°C
Melting Point 216°C
Physical Form Resublimed Crystalline
Quantity 25 g
Assay Percent Range 99.99%, metal basis
Solubility Information Soluble in water (reacts). Also soluble in ethanol,ether and carbon tetrachloride.
Formula Weight 358.21
Sensitivity Moisture sensitive
Density 3.68
Chemical Name or Material Tantalum(V) Chloride, Ampouled under argon

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP07L039 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number010880.14,010880.06,010880.22
Lot NumberU06K024 Certificate TypeCertificate of Analysis Date25/09/2024 Catalog Number010880.14,010880.06,010880.22
Lot NumberM02K039 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog Number010880.14,010880.06,010880.22
Lot NumberZ06J036 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog Number010880.14,010880.06,010880.22
Lot NumberU31J053 Certificate TypeCertificate of Analysis Date22/09/2023 Catalog Number010880.14,010880.06,010880.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.