missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ TAP Growth Media, optimized for Chlamydomonas culture
TAP Growth Media, optimized for Chlamydomonas culture
Brand: Gibco™ A1379802
297.23 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.45000kg
Description
Gibco™ TAP Media is optimized for the growth and maintenance of Chlamydomonas reinhardtii. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.Each Gibco™ TAP Media comes in the award winning Gibco™ bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helping you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Chlamydomonas culture, results in better overall efficiency and more robust data (Fig. 1, 2).
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
Specifications
Gibco™ TAP Growth Media contains: • 6 L Gibco™ TAP Media — Store at 4 °C |
|
Liquid | |
None (Ready-to-Use) |
1 X | |
Growth Medium |