missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Taq DNA Polymerase (cloned)
For PCR and other procedures requiring DNA polymerase activity at elevated temperatures
Brand: Cytiva 27-0798-04
92.41 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
One unit incorporates 10nmol of total nucleotides into acid-insoluble material in 30 min. at 70°C in a total volume of 50μL.
- Purity: Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities
- Concentration: 5000 units/mL
- Functional testing: Tested in PCR
- Storgae Buffer: 50mM Tris-HCl (pH 7.5), 5mM DTT, 0.1mM EDTA, 50% glycerol, stabilizers
Specifications
100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C | |
Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities. | |
Standard PCR |
7.5 | |
250U | |
5000U/mL |
Safety and Handling
Product Identifier
Hazard Statement
Supplemental information
- Taq DNA Polymerase (cloned), 250 units
Hazard Statement
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-1.6 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 1.6% of components with unknown hazards to the aquatic environment.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction