missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Taq DNA Polymerase (cloned)
For PCR and other procedures requiring DNA polymerase activity at elevated temperatures
£97.27 - £530.00
Specifications
Content And Storage | 100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C |
---|---|
pH | 7.5 |
Purity or Quality Grade | Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities. |
For Use With (Application) | Standard PCR |
Concentration | 5000U/mL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10371425
|
Cytiva
27-0798-04 |
250U |
£97.27
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10764237
|
Cytiva
27-0798-05 |
4 x 250U | |||||||
10629275
|
Cytiva
27-0798-06 |
10 x 250U |
£530.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
One unit incorporates 10nmol of total nucleotides into acid-insoluble material in 30 min. at 70°C in a total volume of 50μL.
- Purity: Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities
- Concentration: 5000 units/mL
- Functional testing: Tested in PCR
- Storgae Buffer: 50mM Tris-HCl (pH 7.5), 5mM DTT, 0.1mM EDTA, 50% glycerol, stabilizers
Specifications
100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C | |
Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities. | |
5000U/mL |
7.5 | |
Standard PCR |
Safety and Handling
Product Identifier
Hazard Statement
Supplemental information
- Taq DNA Polymerase (cloned), 250 units
Hazard Statement
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-1.6 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 1.6% of components with unknown hazards to the aquatic environment.