missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Taq DNA Polymerase (cloned)

For PCR and other procedures requiring DNA polymerase activity at elevated temperatures

Brand:  Cytiva 27-0798-05

 View more versions of this product

Product Code. 10764237

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

One unit incorporates 10nmol of total nucleotides into acid-insoluble material in 30 min. at 70°C in a total volume of 50μL.

  • Purity: Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities
  • Concentration: 5000 units/mL
  • Functional testing: Tested in PCR
  • Storgae Buffer: 50mM Tris-HCl (pH 7.5), 5mM DTT, 0.1mM EDTA, 50% glycerol, stabilizers

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl. Store at -20°C
Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities.
Standard PCR
7.5
4 x 250U
5000U/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers