Learn More
MP Biomedicals™ Taq Pol Bulk (15U/μL)
Supplied with standard reaction buffer
Brand: MP Biomedicals™ 11EPTQC200
Code : 50
Additional Details : Weight : 1.00000kg
Includes: Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2
Description
Provided with QBiogene 10X Incubation PCR Buffer which is suitable for broad range of reaction conditions.
Specifications
2000U | |
Standard PCR | |
15U/μL | |
Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2 |
Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.