missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Taq Pol Bulk (15U/μL)
Supplied with standard reaction buffer
£364.64 - £1031.38
Specifications
For Use With (Application) | Standard PCR |
---|---|
Concentration | 15U/μL |
Includes | Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467020
|
MP Biomedicals™
11EPTQC060 |
600U |
£364.64
600 units |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11477020
|
MP Biomedicals™
11EPTQC200 |
2000U |
£1031.38
2000 units |
Estimated Shipment: 12-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Provided with QBiogene 10X Incubation PCR Buffer which is suitable for broad range of reaction conditions.
Specifications
Standard PCR | |
15U/μL | |
Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2 |
Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.