missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Non-Coding RNA Assay
TaqMan™ Non-Coding RNA Assay
Marque: Applied Biosystems™ 4441122
Informations supplémentaires : Weight : 0.03500kg
Description
- TaqMan™ Non-coding RNA Assays provide a highly reliable way to measure expression of long non-coding transcripts.
- These assays are based on TaqMan™ Assays technology to help provide the high specificity and sensitivity you need for quantification and validation studies.
Spécification
Store in freezer (-5° to -30°C). | |
Extra small scale | |
Liquid | |
TaqMan™ |
For Research Use Only. Not for use in diagnostic procedures.