missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Non-Coding RNA Assay
TaqMan™ Non-Coding RNA Assay
£138.00 - £255.00
Specifications
Content And Storage | Store in freezer (-5° to -30°C). |
---|---|
Form | Liquid |
Product Line | TaqMan™ |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
15534334
|
Applied Biosystems™
4441120 |
Small scale |
£255.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
15544334
|
Applied Biosystems™
4441122 |
Extra small scale |
£138.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
|||||
Description
- TaqMan™ Non-coding RNA Assays provide a highly reliable way to measure expression of long non-coding transcripts.
- These assays are based on TaqMan™ Assays technology to help provide the high specificity and sensitivity you need for quantification and validation studies.
Specifications
Store in freezer (-5° to -30°C). | |
Liquid | |
TaqMan™ |
For Research Use Only. Not for use in diagnostic procedures.