missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Protein Assays Core Reagents Base Kit

TaqMan™ Protein Assays Core Reagents Base Kit

Brand:  Applied Biosystems™ 4405460

226.95 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11521306

  • £445.00 / 100 reactions
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TaqMan™ Protein Assays Core Reagents Base Kit contains the reagents required to perform the ligation and PCR amplification steps of TaqMan™ protein expression experiments. The Universal PCR Assay contains optimized concentrations of TaqMan™ probe and oligonucleotide primers (forward and reverse) that are designed to amplify a region of the protease-treated ligation product for detection on an Applied Biosystems real-time PCR system.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, AB 7900 HT 384-Well System, StepOne™, Fast Mode, StepOnePlus™, Fast Mode
Proximity Ligation Assay
Primer-Probe Detection
Reagents for Protein Expression
Cells
TaqMan™
Fast
Not Target-Specific
TaqMan™
The TaqMan™ Protein Assays Core Reagents Base Kit contains:

DNA Ligase, 500X (1 × 10 μL)
Ligase Dilution Buffer, 1X (2 × 1.5 mL)
Ligation Reaction Buffer, 20X (1 × 0.7 mL)
Phosphate Buffered Saline (PBS), 1X, pH 7.4 (1 × 1 mL)
Protease, 100X (1 × 10 μL)
Universal PCR Assay, 20X (1 × 120 μL)
Store all components at -15 to -25°C.
FAM
100 Reactions
100 reactions
qPCR Protein Quantification
Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.