missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Protein Assays Core Reagents Base Kit
TaqMan™ Protein Assays Core Reagents Base Kit
£445.00 - £1342.00
Specifications
Product Line | TaqMan™ |
---|---|
Detection Method | Primer-Probe Detection |
Label or Dye | FAM |
Primer Probe Compatibility | TaqMan™ |
PCR Method | qPCR Protein Quantification |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
11521306
|
Applied Biosystems™
4405460 |
100 reactions |
£445.00
100 reactions |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
12047799
|
Applied Biosystems™
4448592 |
500 |
£1342.00
500 reactions |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
TaqMan™ Protein Assays Core Reagents Base Kit contains the reagents required to perform the ligation and PCR amplification steps of TaqMan™ protein expression experiments. The Universal PCR Assay contains optimized concentrations of TaqMan™ probe and oligonucleotide primers (forward and reverse) that are designed to amplify a region of the protease-treated ligation product for detection on an Applied Biosystems real-time PCR system.Specifications
TaqMan™ | |
FAM | |
qPCR Protein Quantification | |
Wet Ice |
Primer-Probe Detection | |
TaqMan™ | |
Fast | |
Not Target-Specific |
For Research Use Only. Not for use in diagnostic procedures.