missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ TaqMan™ Protein Assays Core Reagents Base Kit
TaqMan™ Protein Assays Core Reagents Base Kit
Brand: Applied Biosystems™ 4448592
684.42 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
TaqMan™ Protein Assays Core Reagents Base Kit contains the reagents required to perform the ligation and PCR amplification steps of TaqMan™ protein expression experiments. The Universal PCR Assay contains optimized concentrations of TaqMan™ probe and oligonucleotide primers (forward and reverse) that are designed to amplify a region of the protease-treated ligation product for detection on an Applied Biosystems real-time PCR system.Specifications
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, StepOnePlus™, Fast Mode | |
Real Time PCR (qPCR), Stem Cell Analysis | |
Primer-Probe Detection | |
Protein Assays Core Reagents Base Kit | |
No | |
ROX (Pre-mixed) | |
TaqMan™ | |
Fast | |
Not Target-Specific |
TaqMan™ | |
The TaqMan™ Protein Assays Core Reagents Base Kit contains: DNA Ligase, 500X (1 × 50 μL) Ligase Dilution Buffer, 1X (1 × 15 mL) Ligation Reaction Buffer, 20X (1 × 4 mL) Phosphate Buffered Saline (PBS), 1X, pH 7.4 (1 × 5 mL) Protease, 100X (1 × 50 μL) Universal PCR Assay, 20X (1 × 600 μL) Store all components at -15 to -25°C. |
|
FAM | |
Tissue, Cells | |
500 | |
AmpliTaq™ Fast DNA Polymerase | |
qPCR Protein Quantification | |
Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.