missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCEANC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-13419
This item is not returnable.
View return policy
Description
TCEANC2 Polyclonal specifically detects TCEANC2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TCEANC2 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
C1orf83, chromosome 1 open reading frame 83, FLJ32112, FLJ39169, RP4-758J24.3, transcription elongation factor A (SII) N-terminal and central domaincontaining 2 | |
Rabbit | |
Affinity Purified | |
RUO | |
127428 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TCEANC2 | |
This antibody was developed against a recombinant protein corresponding to the amino acids: TMLELPDQTKENLVEALQELKKKIPSREVLKSTRIGHTVNKMRKHSDSEVASLAREVYTEWKTFTEKHSNRPSIEV | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only