Learn More
TCR gamma/delta Monoclonal Antibody (UC7-13D5), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand:  Affymetrix eBioscience 16-5811-025
 View more versions of this product
Product Code. 17101312
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
The UC7-13D5 MAb reacts with the mouse gamma delta TCR complex. The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection.
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry, Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TCR gamma/delta
Monoclonal
1 mg/mL
TBS with no preservative; pH 8.0
T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@
Affinity chromatography
RUO
110066, 110067
4° C
Liquid
Flow Cytometry, Functional Assay
UC7-13D5
Functional Grade
Tcrg
Armenian Hamster
25 mg