Learn More
TCR gamma/delta Monoclonal Antibody (UC7-13D5), Functional Grade, eBioscience™, Invitrogen™
Armenian Hamster Monoclonal Antibody
£2595.00 - £12950.00
Specifications
Antigen | TCR gamma/delta |
---|---|
Clone | UC7-13D5 |
Concentration | 1 mg/mL |
Applications | Flow Cytometry, Functional Assay |
Classification | Monoclonal |
Description
The UC7-13D5 MAb reacts with the mouse gamma delta TCR complex. The gamma delta TCR is expressed by a small subset of T cells in the thymus, peripheral lymphoid tissues, intestinal epithelium, and epidermis. The exact specificity, ligand and function of gamma delta TCR-bearing T cells are not yet fully understood; it is suggested that these cells recognize bacterial ligands and some tumor cells in the context of MHC class I-like gene products and play a role in regulation of the immune response and during bacterial infection.
TCR gamma/delta Monoclonal antibody specifically detects TCR gamma/delta in Mouse samples. It is validated for Flow Cytometry, Functional AssaySpecifications
TCR gamma/delta | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
TBS with no preservative; pH 8.0 | |
110066, 110067 | |
Primary | |
4° C |
UC7-13D5 | |
Flow Cytometry, Functional Assay | |
Functional Grade | |
Armenian Hamster | |
Mouse | |
T cell receptor gamma locus; T3/TCR complex; TCR delta gamma; TCR gamma/delta; TCR-CD3 complex; Tcrg; Trg; Trg@ | |
IgG | |
Affinity chromatography | |
Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.