Learn More
TCR V beta 5.1/5.2 Monoclonal Antibody (MR9-4), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£115.00 - £286.00
Specifications
Antigen | TCR V beta 5.1/5.2 |
---|---|
Clone | MR9-4 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15598126
|
Invitrogen
17-5796-80 |
25 μg |
£115.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15508136
|
Invitrogen
17-5796-82 |
100 μg |
£286.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This MR9-4 monoclonal antibody reacts with the mouse T cell receptor (TCR) V beta 5.1/5.2 chain. Composed of an alpha and beta chain, TCR specificity is typically determined by Va, Ja, Vb, Db, and Jb gene rearrangement. The MR9-4 antibody recognizes the V beta 5.1 and 5.2 chains on T cells from mouse strains with the b haplotype of the Tcrb gene, including C57BL/6, B10, and C58/J. V beta 5.1 and 5.2 are deleted in mouse strains that express MHC Class II I-E, including Balb/c, C3H, and DBA/2. Reports also indicate that V beta 5+ T cells recognize the OVA peptide. Applications Reported: This MR9-4 antibody has been reported for use in flow cytometric analysis. Applications Tested: This MR9-4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
TCR V beta 5.1/5.2 Monoclonal antibody specifically detects TCR V beta 5.1/5.2 in Mouse samples. It is validated for Flow Cytometry.Specifications
TCR V beta 5.1/5.2 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
MR9-4 | |
Flow Cytometry | |
APC | |
Mouse | |
Mouse | |
TCR V beta5.1; TCR V beta5.2; TCRV beta 5.1; TCRV beta 5.2; TCRV beta5.1; TCRV beta5.2; Vb5.1; Vb5.2; Vbeta5.1; Vbeta5.2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
For Research Use Only.