missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ WBU-200 Series Unheated Water Bath

This WBU-200 Series Unheated Water Bath features a corrosion resistant stainless steel inner for easy cleaning.

Brand:  Cole-Parmer™ 16104-53

375.60 GBP valid until 2025-12-31
Use promo code "24307" to get your promotional price.


 View more versions of this product

Product Code. 12777919

  • £489.00 / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of durable stainless steel.
  • Rugged splash-proof case.
  • Integrated carrying handle for added safety.
  • Designed to be used with a clip-on Tempette or Tempunit™ thermoregulator (sold separately), these water baths incorporate carrying handles for added safety.
  • All water baths have stoved enamelled steel outer cases and are supplied with bridge mounting plate to hold the thermoregulator.
Specifications

Specifications

2.1 gal.
Waterbath
Clip-on Tempette
8 to 6 L
200°C
240 mm
300 mm
5.9 in.
150 mm
65 mm
Stainless Steel
8 L
Unheated Water Bath
Stainless Steel
265 mm
325 mm
6.8 in.
172 mm
2.6 in.
5.5 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.