missing translation for 'onlineSavingsMsg'
Learn More

Technical pH Buffer Solution, pH 9.21, Mettler Toledo™
GREENER_CHOICE

Product Code. 12631306
Click to view available options
Quantity:
6 x 250 mL
Packaging:
6 x 250mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Traceable to the primary standards of reference materials selected by NIST.

  • pH 9.21
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
pH 9.21
Physical Form Liquid
Quantity 6 x 250 mL
Solution Type pH Buffer
Odor None
Packaging 6 x 250mL
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.