missing translation for 'onlineSavingsMsg'
Learn More
Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals Product Code.: 11357787

Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals

Product Code. 11357787
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11357787

Brand: Thermo Scientific Alfa Aesar 014197.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Telluric acid dihydrate is used in the preparation of tellurium nanorods through hydrazine-driven reduction, which finds application in solar cells, thermoelectric and topological insulators. It is also involved in the preparation of tellurium metal by reacting with the solution of potassium thiocyanate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13451-14-4
Molecular Formula H6O6Te
Molecular Weight (g/mol) 229.64
MDL Number MFCD00150387
InChI Key VCYUGZHEVVXOPD-UHFFFAOYSA-N
Synonym telluric acid dihydrate, telluricacidwhitepowder, telluric acid-te

Specifications

Boiling Point 160°C (decomposition)
Melting Point 136°C
Color White
Physical Form Crystalline
Quantity 50 g
Assay Percent Range >99%
Solubility Information Soluble in water.
Formula Weight 229.64 (193.61 Anhydrous)
Sensitivity Hygroscopic
Density 3.158
Chemical Name or Material Telluric acid dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberC25U041 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014197.18,014197.09
Lot NumberD26L48 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014197.18,014197.09
Lot NumberA31Q15 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014197.18,014197.09
Lot NumberH07N25 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014197.18,014197.09
Lot NumberC02R023 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number014197.18,014197.09
compliance-icons
Product Identifier
  • Telluric acid dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P362-Take off contaminated clothing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Telluric acid dihydrate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.