missing translation for 'onlineSavingsMsg'
Learn More
Tellurium(IV) oxide, Puratronic, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11377057

Tellurium(IV) oxide, Puratronic, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11377057
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11377057

Brand: Thermo Scientific Alfa Aesar 010882.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-07-3
Molecular Formula O2Te
Molecular Weight (g/mol) 159.60
MDL Number MFCD00011263
InChI Key LAJZODKXOMJMPK-UHFFFAOYSA-N
Synonym tellurium iv oxide, tellurium oxide teo2, tellurite teo2, teo2, unii-397e9rke83, oxotellane oxide, telluride oxide, 6-chlorobutyric acid, acmc-1bglqShow MoreShow Less
PubChem CID 62638
SMILES O=[Te]=O

Specifications

Melting Point 733°C
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.9995%, metal basis
Solubility Information Soluble in strong acids and alkali. Insoluble in water.
IUPAC Name (oxo-λ⁴-tellanylidene)oxidane
Formula Weight 159.6
Density 5.8
Chemical Name or Material Tellurium(IV) Oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX03K027 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number010882.14,010882.36,010882.22
Lot NumberU12F005 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number010882.14,010882.36,010882.22
Lot NumberT17F029 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number010882.14,010882.36,010882.22
Lot NumberQ07I006 Certificate TypeCertificate of Analysis Date20/04/2022 Catalog Number010882.14,010882.36,010882.22
Lot NumberM04I017 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number010882.14,010882.36,010882.22
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity on or via lactation
  • Reproductive toxicity Category 1B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H362-May cause harm to breast-fed children.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tellurium(IV) oxide, Puratronic, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.