Learn More
Tuttnauer Temperature Hold Cycle
Special program with programmable holding temperature at the end of the cycle. This will prevent that media will cool down completely. Must specify the cycle parameters, the holding temperature after the end of sterilization and the length of time to hold the temperature.
Brand:  Tuttnauer ADD222-0460
Product Code. 17829538
  • £1480.00 / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Special program with programmable holding temperature at the end of the cycle.
  • This will prevent that media will cool down completely.
  • Must specify the cycle parameters, the holding temperature after the end of sterilization and the length of time to hold the temperature.
Specifications
Specifications
Software
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tuttnauer Temperature Hold Cycle > Temperature Hold Cycle