missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Temperature Restriction for UF30
Optional temperature restriction between 60°C and 250°C.
Brand: Memmert™ UF30A8
This item is not returnable.
View return policy
Description
- Accessory
- 1 Choice
- Temperatures: 60°C, 70°C, 80°C, 95°C, 100°C, 120°C, 160°C, 180°C, 200°C, or 250°C
Specifications
UF30 Oven | |
Temperature Restriction |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Temperature Restriction for UF30 > For Use With: UF30
Spot an opportunity for improvement?Share a Content Correction