Learn More
Dostmann Electronic™ Temperature Sensor, Thermocouple
Dostmann Electronic™ Temperature Sensor, Thermocouple is a plug and cable type temperature sensor suitable for variety of temperature measurements demands.
Brand:  Dostmann Electronic™ 6010-0088-6
 View more versions of this product
Product Code. 11826850
  • £48.00 / Each
Explore more special offers
Description
Description
  • Type K according to EN 60584-1
  • NiCr-Ni
  • Mineral-insulated
Specifications
Specifications
3 mm
Temperature Sensor
-40°C to +480°C
6000 mm
Steel braiding with thermocouple strip
P410 / TC301 / TC 309 / Mini-K