Learn More
Cytiva TempliPhi™ Sequence Resolver Kit
Uses modified nucleotides in isothermal rolling circle amplification (RCA) to improve results with difficult to sequence templates
Brand: Cytiva 28-9035-31
1493.37 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GE
Additional Details : CAS Number : 7732-18-5 Weight : 0.02700kg
Includes: Sample buffer, Reaction buffer and Enzyme mix
Description
- Cultureless preparation from bacterial colonies, glycerol stock or purified DNA, saves time and reagents
- Requires 20 minutes hands-on time
- Excellent sequence finishing performance resolving sequencing stop and compression issues associated with difficult templates
- Improve difficult template sequencing success with up to 820 bp Phred 20 read length
- For use directly in cycle sequencing reactions without purification
- Available in sizes of 20, 50 and 500 reactions
Specifications
Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°CThaw components on ice and maintain at 0°C to 4°C during handling | |
Amplification of difficult templates for successful DNA sequencing | |
200 | |
18 h |
illustra TempliPhi Sequence Resolver Kit uses the highly processive, strand-displacing Phi29 DNA polymerase and modified nucleotides to amplify circular templates for subsequent sequencing. | |
1 x 900μL Sample Buffer, 1 x 900μL Reaction Buffer, 1 x 100μL Enzyme mix | |
illustra TempliPhi Sequence Resolver Kit |
Safety and Handling
- TempliPhi Sequence Resolver Kit, 200 reactions
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : glycerol