missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva TempliPhi™ Sequence Resolver Kit
Uses modified nucleotides in isothermal rolling circle amplification (RCA) to improve results with difficult to sequence templates
£377.00 - £1907.00
Specifications
Content And Storage | Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°CThaw components on ice and maintain at 0°C to 4°C during handling |
---|---|
Description | illustra TempliPhi Sequence Resolver Kit uses the highly processive, strand-displacing Phi29 DNA polymerase and modified nucleotides to amplify circular templates for subsequent sequencing. |
For Use With (Application) | Amplification of difficult templates for successful DNA sequencing |
Product Type | illustra TempliPhi Sequence Resolver Kit |
Test Time | 18 h |
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | No. of Reactions | Price | Quantity & Availability | ||||
10308259
|
Cytiva
28-9035-31 |
1 x 900μL Sample Buffer, 1 x 900μL Reaction Buffer, 1 x 100μL Enzyme mix | 200 |
£1907.00
1 set |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11339941
|
Cytiva
28-9035-29 |
1 x 90μL Sample Buffer, 1 x 90μL Reaction Buffer, 1 x 90μL Enzyme mix | 20 |
£377.00
1 set |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Cultureless preparation from bacterial colonies, glycerol stock or purified DNA, saves time and reagents
- Requires 20 minutes hands-on time
- Excellent sequence finishing performance resolving sequencing stop and compression issues associated with difficult templates
- Improve difficult template sequencing success with up to 820 bp Phred 20 read length
- For use directly in cycle sequencing reactions without purification
- Available in sizes of 20, 50 and 500 reactions
Specifications
Store the kit at -70°C. The enzyme mix must be stored at -70°C; all other components may be stored at -20°CThaw components on ice and maintain at 0°C to 4°C during handling | |
Amplification of difficult templates for successful DNA sequencing | |
18 h |
illustra TempliPhi Sequence Resolver Kit uses the highly processive, strand-displacing Phi29 DNA polymerase and modified nucleotides to amplify circular templates for subsequent sequencing. | |
illustra TempliPhi Sequence Resolver Kit |