missing translation for 'onlineSavingsMsg'
Learn More
Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11326248

Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11326248
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11326248

Brand: Thermo Scientific Alfa Aesar 011207.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Terbium(III,IV) oxide acts as a precursor for the preparation of other terbium compounds. It is used as a redox catalyst in reactions involving oxygen. At high temperatures, it can liberate oxygen. It finds applications in the manufacture of glass, optical and ceramic. Nanoparticles of terbium oxide are used as analytical reagents for the determination of drugs in food.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12037-01-3
Molecular Formula O7Tb4
Molecular Weight (g/mol) 747.69
MDL Number MFCD00083151
InChI Key UWZVBPNIJCKCMC-UHFFFAOYSA-N
IUPAC Name tetraoxotetraterboxane
SMILES O=[Tb]O[Tb](=O)O[Tb](=O)O[Tb]=O

Specifications

Melting Point 2340°C
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.9% (REO)
Solubility Information Soluble in dilute acids. Insoluble in water.
Formula Weight 747.69
Chemical Name or Material Terbium(III, IV) oxide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ11J056 Certificate TypeCertificate of Analysis Date08/01/2024 Catalog Number011207.30,011207.A1,011207.18,011207.09
Lot NumberU15J026 Certificate TypeCertificate of Analysis Date14/09/2023 Catalog Number011207.30,011207.A1,011207.18,011207.09
Lot Number61301170 Certificate TypeCertificate of Analysis Date23/11/2022 Catalog Number011207.30,011207.A1,011207.18,011207.09
Lot Number61200598 Certificate TypeCertificate of Analysis Date23/11/2022 Catalog Number011207.30,011207.A1,011207.18,011207.09
Lot Number61301170 Certificate TypeCertificate of Analysis Date22/11/2022 Catalog Number011207.30,011207.A1,011207.18,011207.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Terbium(III,IV) oxide, REacton™, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.