missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Terg-A-Zyme™ Cleaner

Use this superior cleaners on glass, metal, plastic, rubber and other hard materials in your critical cleaning processes. Terg-A-Zyme™ Cleaner rinses off completely with no interfering residue.

Brand:  Cole-Parmer™ 17777-05

67.95 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.



Product Code. 11787168

  • £72.70 / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Use this superior cleaners on glass, metal, plastic, rubber and other hard materials in your critical cleaning processes. Terg-A-Zyme™ Cleaner rinses off completely with no interfering residue.

White powder cleaner is safe, odorless, and FDA compliant.
  • Terg-A-Zyme cleaner combines the original Alconox; power with protein enzymes.
  • ALCONOX and TERG-A-ZYME are registered trademarks of Alconox Inc.
Specifications

Specifications

Terg-A-Zyme Cleaner
FDA compliant
For Protein Removal
1 Ea.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium tripolyphosphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Terg-A-Zyme™ Cleaner > Weight: 1.8kg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.