missing translation for 'onlineSavingsMsg'
Learn More
tert-Butanol, Thermo Scientific Chemicals Product Code.: 10667464

tert-Butanol, Thermo Scientific Chemicals

Product Code. 10667464
1 L, Plastic Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10667464

Brand: Thermo Scientific Acros 390690010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-65-0
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004464
InChI Key DKGAVHZHDRPRBM-UHFFFAOYSA-N
Synonym tert-butanol, tert-butyl alcohol, 2-methyl-2-propanol, t-butanol, t-butyl hydroxide, 1,1-dimethylethanol, trimethylcarbinol, trimethyl methanol, 2-propanol, 2-methyl, t-butyl alcoholShow MoreShow Less
PubChem CID 6386
ChEBI CHEBI:45895
IUPAC Name 2-methylpropan-2-ol
SMILES CC(C)(C)O

Specifications

Melting Point 25.0°C to 25.5°C
Density 0.7800g/mL
Boiling Point 83.0°C
Quantity 1 L
Flash Point 11°C
Infrared Spectrum Authentic
Packaging Plastic Bottle
Physical Form Liquid
Refractive Index 1.3860 to 1.3880
Residue after Evaporation 0.001% max.
Specific Gravity 0.78
Titratable Acid 0.0005 meq/g max.
Linear Formula (CH3)3COH
UN Number 1120
Viscosity 3.35 mPa.s (30°C)
Water 0.1% max. (K.F.)
Merck Index 15, 1544
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol, ether, acetone
Formula Weight 74.12
Percent Purity ≥99.5%
Grade ACS Reagent
Chemical Name or Material tert-Butanol, For analysis ACS
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464391 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog Number390690010,390690025
Lot NumberA0455525 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number390690010,390690025
Lot NumberA0451128 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number390690010,390690025
Lot NumberA0446258 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number390690010,390690025
Lot NumberA0438565 Certificate TypeCertificate of Analysis Date17/02/2022 Catalog Number390690010,390690025
compliance-icons compliance-icons
Product Identifier
  • tert-Butanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butanol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.