Learn More
tert-Butyl 2,2,2-trichloroacetimidate, 97%, Thermo Scientific Chemicals
£26.90 - £147.00
Chemical Identifiers
CAS | 98946-18-0 |
---|---|
Molecular Formula | C6H10Cl3NO |
Molecular Weight (g/mol) | 218.50 |
MDL Number | MFCD00077410 |
InChI Key | CQXDYHPBXDZWBA-UHFFFAOYSA-N |
PubChem CID | 2734700 |
IUPAC Name | tert-butyl 2,2,2-trichloroethanimidate |
SMILES | CC(C)(C)OC(=N)C(Cl)(Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483950
|
Thermo Scientific Alfa Aesar
B22039.03 |
1 g |
£26.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11479527
|
Thermo Scientific Alfa Aesar
B22039.06 |
5 g |
£39.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489527
|
Thermo Scientific Alfa Aesar
B22039.14 |
25 g |
£147.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
tert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl 2,2,2-trichloroacetimidate is used to produce di-tert-butyl peroxide at temperature of -5°C. It may be used in the synthesis of N-α-Fmoc-phospho(1-nitrophenylethyl-2-cyanoethyl)-L-serine, caged building block. It may be used in the conversion of alcohols and carboxylic acids to their respective ethers and esters.
Solubility
Soluble in organic solvents, cyclo hexane.
Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.
Chemical Identifiers
98946-18-0 | |
218.50 | |
CQXDYHPBXDZWBA-UHFFFAOYSA-N | |
tert-butyl 2,2,2-trichloroethanimidate |
C6H10Cl3NO | |
MFCD00077410 | |
2734700 | |
CC(C)(C)OC(=N)C(Cl)(Cl)Cl |
Specifications
98946-18-0 | |
1.222 | |
55°C (131°F) | |
C6H10Cl3NO | |
MFCD00077410 | |
1770049 | |
tert-butyl 2,2,2-trichloroacetimidate, tert-butyl trichloroacetimidate, tert-butyl-2,2,2-trichloroacetimidate, tert-butyl 2,2,2-trichloroethanecarboximidate, 2,2,2-trichloroacetimidic acid tert-butyl ester, t-butyl trichloroacetimidate, t-butyl 2,2,2-trichloroacetimidate, tert-butyl2,2,2-trichloroacetimidate, ethanimidic acid, 2,2,2-trichloro-, 1,1-dimethylethyl ester, 1-tert-butoxy-2,2,2-trichloroethanimine | |
CQXDYHPBXDZWBA-UHFFFAOYSA-N | |
tert-butyl 2,2,2-trichloroethanimidate | |
2734700 | |
97% |
20°C to 22°C | |
64°C to 66°C (15 mmHg) | |
0.97 | |
1.457 | |
UN1993 | |
Moisture sensitive | |
Soluble in organic solvents,cyclo hexane. | |
CC(C)(C)OC(=N)C(Cl)(Cl)Cl | |
218.50 | |
218.51 | |
tert-Butyl 2,2,2-trichloroacetimidate |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
missing translation for 'storageNote' : Moisture Sensitive
Recommended Storage : Keep cold
RUO – Research Use Only