missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Thermo Scientific™

£188.06 - £634.83

Chemical Identifiers

CAS 844891-13-0
Molecular Formula C11H16N2O3
Molecular Weight (g/mol) 224.26
MDL Number MFCD06659076
InChI Key AZJWODKWFMBMCM-UHFFFAOYSA-N
PubChem CID 2795543
IUPAC Name tert-butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate
SMILES CC1=NN(C(=O)OC(C)(C)C)C(C)=C1C=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10402273
SDS View Documents
Thermo Scientific Maybridge
CC31904DA
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10413214
SDS View Documents
Thermo Scientific Maybridge
CC31904DE
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.