Learn More
tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals
£29.50 - £86.80
Chemical Identifiers
CAS | 614-45-9 |
---|---|
Molecular Weight (g/mol) | 194.23 |
MDL Number | MFCD00008802 |
InChI Key | GJBRNHKUVLOCEB-UHFFFAOYSA-N |
Synonym | tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan |
PubChem CID | 11966 |
IUPAC Name | tert-butyl benzenecarboperoxoate |
SMILES | CC(C)(C)OOC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10636572
|
Thermo Scientific Acros
170142500 |
250 g | Plastic bottle |
£29.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10381424
|
Thermo Scientific Acros
170140010 |
1 kg | Plastic bottle |
£86.80
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsChemical Identifiers
614-45-9 | |
MFCD00008802 | |
tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan | |
tert-butyl benzenecarboperoxoate |
194.23 | |
GJBRNHKUVLOCEB-UHFFFAOYSA-N | |
11966 | |
CC(C)(C)OOC(=O)C1=CC=CC=C1 |
Specifications
614-45-9 | |
Yellow | |
75.0°C to 76.0°C (0.2 mmHg) | |
Authentic | |
C11H14O3 | |
1.4980 to 1.5000 | |
09, IV, 715 | |
1.04 | |
Solubility in water: immiscible. Other solubilities: soluble in alcohols,esters,ethers and ketones | |
CC(C)(C)OOC(=O)C1=CC=CC=C1 | |
194.23 | |
6 mPa.s (20°C) | |
98% | |
tert-Butyl peroxybenzoate, 98% |
8.0°C | |
1.0400g/mL | |
93°C | |
97.5% min. (GC) | |
C6H5CO2OC(CH3)3 | |
MFCD00008802 | |
01,98; 02,54; 04,66; 07,49; 09,90; 13,58 | |
tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan | |
GJBRNHKUVLOCEB-UHFFFAOYSA-N | |
tert-butyl benzenecarboperoxoate | |
11966 | |
194.23 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if inhaled.
Heating may cause a fire.
May cause an allergic skin reaction.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from cl
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 210-382-2
missing translation for 'rtecsNumber' : SD9450000
missing translation for 'tsca' : TSCA
- tert-Butyl peroxybenzoate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Organic peroxide Type C & D
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H242-Heating may cause a fire.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P234-Keep only in original container.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only