missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals Product Code.: 10636572

tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 10636572
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10636572

Brand: Thermo Scientific Acros 170142500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-45-9
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008802
InChI Key GJBRNHKUVLOCEB-UHFFFAOYSA-N
Synonym tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoanShow MoreShow Less
PubChem CID 11966
IUPAC Name tert-butyl benzenecarboperoxoate
SMILES CC(C)(C)OOC(=O)C1=CC=CC=C1

Specifications

Melting Point 8.0°C
Color Yellow
Density 1.0400g/mL
Boiling Point 75.0°C to 76.0°C (0.2 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Molecular Formula C11H14O3
Linear Formula C6H5CO2OC(CH3)3
Refractive Index 1.4980 to 1.5000
Quantity 250 g
Beilstein 09, IV, 715
Fieser 01,98; 02,54; 04,66; 07,49; 09,90; 13,58
Specific Gravity 1.04
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohols,esters,ethers and ketones
Viscosity 6 mPa.s (20°C)
Formula Weight 194.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material tert-Butyl peroxybenzoate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467010 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number170142500,170140010
Lot NumberA0438826 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number170142500,170140010
Lot NumberA0403583 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number170142500,170140010
Lot NumberA0435873 Certificate TypeCertificate of Analysis Date01/11/2021 Catalog Number170142500,170140010
Lot NumberA0428184 Certificate TypeCertificate of Analysis Date16/03/2021 Catalog Number170142500,170140010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • tert-Butyl peroxybenzoate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Organic peroxide Type C & D
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H242-Heating may cause a fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.