missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylchlorodiphenylsilane, 98%, Thermo Scientific Chemicals

£34.27 - £452.00

Chemical Identifiers

CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.86
MDL Number MFCD00000497
InChI Key MHYGQXWCZAYSLJ-UHFFFAOYSA-N
Synonym tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
PubChem CID 94078
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10593741
View Documents
Thermo Scientific Acros
187500100
10 g Glass bottle
£34.27
10g
Please to purchase this item. Need a web account? Register with us today!
10467560
View Documents
Thermo Scientific Acros
187500500
50 g Glass bottle
£108.54
50g
Please to purchase this item. Need a web account? Register with us today!
10247523
View Documents Promotion Details
Thermo Scientific Acros
187502500
250 g Glass bottle
£452.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

58479-61-1
274.86
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
94078
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
C16H19ClSi
MFCD00000497
tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
tert-butyl-chloro-diphenylsilane

Specifications

58479-61-1
1.0600g/mL
149°C
97.5% min. (GC)
(CH3)3CSi(C6H5)2Cl
MFCD00000497
1.06
Solubility in water: practically insoluble. Other solubilities: soluble in non-protic solvents,e.g. aliphatic,or aromatic (chlorinated) hydrocarbons
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
274.86
16 mPa.s (25°C)
98%
tert-Butylchlorodiphenylsilane
Brown or Colorless to Yellow
135°C (0.7 mmHg)
Authentic
C16H19ClSi
1.5665 to 1.5685
15, 1568
tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
tert-butyl-chloro-diphenylsilane
94078
274.86
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 261-282-

Recommended Storage : May darken during storage

Product Identifier
  • tert-Butylchlorodiphenylsilane

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.