Learn More
Retsch™ Test Sieves with 1.00 to 10.00 mm Mesh Size
Retsch™ Test Sieves consist of a solid stainless steel sieve frame with high stability, resulting in reliable sieving results.
£115.00 - £159.00
Specifications
Certifications/Compliance DIN ISO 3310-1
Material Stainless Steel
Includes Calibration Certificate
Height (Metric) 50 mm
Diameter (Metric) Outer 200 mm
View More Specs
Products 27
Product Code Brand Pore Size Price Quantity & Availability  
Product Code Brand Pore Size Price Quantity & Availability  
15598841
View Documents
Retsch™
60.131.001000
1 mm
£157.00
Each
Estimated Shipment: 11-06-2025
to see stock.
Add to basket
15660389
View Documents
Retsch™
60.131.001120
1.12 mm
£115.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15670389
View Documents
Retsch™
60.131.001180
1.18 mm
£120.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15680389
View Documents
Retsch™
60.131.001250
1.25 mm
£120.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15508851
View Documents
Retsch™
60.131.001400
1.40 mm
£159.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15518851
View Documents
Retsch™
60.131.001600
1.60 mm
£120.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15690389
View Documents
Retsch™
60.131.001700
1.70 mm
£120.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15600399
View Documents
Retsch™
60.131.001800
1.80 mm
£115.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15520171
View Documents
Retsch™
60.131.002000
2.00 mm
£144.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15610399
View Documents
Retsch™
60.131.002240
2.24 mm
£115.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15620399
View Documents
Retsch™
60.131.002360
2.36 mm
£124.00
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
15538851
View Documents
Retsch™
60.131.002500
2.50 mm
£115.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15548851
View Documents
Retsch™
60.131.002800
2.80 mm
£150.80
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15558851
View Documents
Retsch™
60.131.003150
3.15 mm
£151.00
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
15630399
View Documents
Retsch™
60.131.003350
3.35 mm
£141.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15640399
View Documents
Retsch™
60.131.003550
3.55 mm
£141.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15570171
View Documents
Retsch™
60.131.004000
4.00 mm
£151.84
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15650399
View Documents
Retsch™
60.131.004500
4.50 mm
£141.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15568851
View Documents
Retsch™
60.131.004750
4.75 mm
£147.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15540171
View Documents
Retsch™
60.131.005000
5.00 mm
£141.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15660399
View Documents
Retsch™
60.131.005600
5.60 mm
£141.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15578851
View Documents
Retsch™
60.131.006300
6.30 mm
£141.00
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
15670399
View Documents
Retsch™
60.131.006700
6.70 mm
£141.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15598851
View Documents
Retsch™
60.131.008000
8.00 mm
£145.00
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
15680399
View Documents
Retsch™
60.131.009000
9.00 mm
£141.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15690399
View Documents
Retsch™
60.131.009500
9.50 mm
£141.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
15528851
View Documents
Retsch™
60.131.010000
10.00 mm
£157.00
Each
Estimated Shipment: 27-06-2025
to see stock.
Add to basket
Description
Description
  • High quality joining technology guarantees permanently taut wire mesh
  • One-piece cast sieve frame and seamless mesh joint prevent cross contamination
  • Stainless steel sieve frame with high form stability
  • High degree of corrosion resistance and easy cleaning thanks to high-alloy stainless steel
  • Excellent product quality due to extensive optical inspection
  • Maximum stability and optimum sealing when used in sieve stacks thanks to the o-ring which is placed in the recess designed for this purpose
  • Clear and precise labeling of the sieves with full traceability based on individualized laser engraving
Specifications
Specifications
DIN ISO 3310-1
Calibration Certificate
200 mm
Stainless Steel
50 mm
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Retsch™ Test Sieves with 1.00 to 10.00 mm Mesh Size