missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Test Tube Caps
Reduce risk of sample contamination and spillage with a polyethylene constructed cap. Kartell™ Test Tube Caps are specially designed to accommodate both platic and glass test tubes.
Brand: Kartell™ 0065100
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- Constructed of PE
- Designed to fit plastic or glass test tubes
Specifications
CE/IVD | |
Polyethylene | |
16 mm Test Tubes |
14 mm | |
Natural |