missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Test Tube Caps

Reduce risk of sample contamination and spillage with a polyethylene constructed cap. Kartell™ Test Tube Caps are specially designed to accommodate both platic and glass test tubes.

Brand:  Kartell™ 0065700

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10254331

  • £14.15 / Pack of 100
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

  • Constructed of PE
  • Designed to fit plastic or glass test tubes
Specifications

Specifications

CE/IVD
Polyethylene
35 mm Diameter Test Tubes
33 mm
Natural
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers