Learn More
Fisherbrand™ Foam Stands for Vortex Mixer
Securely holds test tubes and vials. Velp Scientifica™ Foam Stands are compatible with Velp Scientifica Vortex Mixers.
Brand:  Fisherbrand™ A00000019
 View more versions of this product
Product Code. 11840992
  • £94.10 / Each
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Black in color
  • Soft foam to prevent breakage
  • Available in different diameters and capacities
Specifications
Specifications
29 mm Tubes
Foam Tube Rack
4
29 mm
CLASSIC, ZX3, WIZARD, ZX4, TX4