missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium benzoate, ca 85%, water ca 10%, Thermo Scientific™

£14.87 - £208.00

Chemical Identifiers

CAS 16436-29-6
Molecular Formula C31H57NO2
Molecular Weight (g/mol) 475.80
MDL Number MFCD00041982
InChI Key PSEQWFPWQRZBOO-UHFFFAOYSA-M
Synonym tetrahexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate, tetrahexylazanium benzoate, tetra-n-hexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate 1:1, tetrahexylazanium, thab, ammonium, tetrahexyl-, benzoate, ammonium, benzoate, tetrahexylazanium,benzoate
PubChem CID 85419
IUPAC Name tetrahexylazanium;benzoate
SMILES [O-]C(=O)C1=CC=CC=C1.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11430878
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B25291.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11440878
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B25291.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11450878
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B25291.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

16436-29-6
475.80
PSEQWFPWQRZBOO-UHFFFAOYSA-M
85419
[O-]C(=O)C1=CC=CC=C1.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
C31H57NO2
MFCD00041982
tetrahexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate, tetrahexylazanium benzoate, tetra-n-hexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate 1:1, tetrahexylazanium, thab, ammonium, tetrahexyl-, benzoate, ammonium, benzoate, tetrahexylazanium,benzoate
tetrahexylazanium;benzoate

Specifications

16436-29-6
MFCD00041982
PSEQWFPWQRZBOO-UHFFFAOYSA-M
tetrahexylazanium;benzoate
85419
∼85%
Tetra-n-hexylammonium benzoate
C31H57NO2
tetrahexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate, tetrahexylazanium benzoate, tetra-n-hexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate 1:1, tetrahexylazanium, thab, ammonium, tetrahexyl-, benzoate, ammonium, benzoate, tetrahexylazanium,benzoate
[O-]C(=O)C1=CC=CC=C1.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC
475.80
456.73
water ≈10%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 240-492-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications