missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraacetylethylenediamine, 90%, Thermo Scientific Chemicals
£33.90 - £102.00
Chemical Identifiers
CAS | 10543-57-4 |
---|---|
Molecular Formula | C10H16N2O4 |
Molecular Weight (g/mol) | 228.25 |
MDL Number | MFCD00014967 |
InChI Key | BGRWYDHXPHLNKA-UHFFFAOYSA-N |
Synonym | tetraacetylethylenediamine, n,n,n',n'-tetraacetylethylenediamine, taed, tetracetylethylenediamine, acetamide, n,n'-1,2-ethanediylbis n-acetyl, n,n'-ethylenebis n-acetylacetamide, unii-p411ed0n2b, n,n'-ethane-1,2-diyl bis n-acetylacetamide, n-acetyl-n-2-diacetylamino ethyl acetamide |
PubChem CID | 66347 |
IUPAC Name | N-acetyl-N-[2-(diacetylamino)ethyl]acetamide |
SMILES | CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10454955
|
Thermo Scientific Acros
344590500 |
50 g | Plastic bottle |
£33.90
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10224951
|
Thermo Scientific Acros
344592500 |
250 g | Plastic bottle |
£102.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10543-57-4 | |
228.25 | |
BGRWYDHXPHLNKA-UHFFFAOYSA-N | |
66347 | |
CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C |
C10H16N2O4 | |
MFCD00014967 | |
tetraacetylethylenediamine, n,n,n',n'-tetraacetylethylenediamine, taed, tetracetylethylenediamine, acetamide, n,n'-1,2-ethanediylbis n-acetyl, n,n'-ethylenebis n-acetylacetamide, unii-p411ed0n2b, n,n'-ethane-1,2-diyl bis n-acetylacetamide, n-acetyl-n-2-diacetylamino ethyl acetamide | |
N-acetyl-N-[2-(diacetylamino)ethyl]acetamide |
Specifications
10543-57-4 | |
White to Beige | |
140°C | |
88% min | |
MFCD00014967 | |
tetraacetylethylenediamine, n,n,n',n'-tetraacetylethylenediamine, taed, tetracetylethylenediamine, acetamide, n,n'-1,2-ethanediylbis n-acetyl, n,n'-ethylenebis n-acetylacetamide, unii-p411ed0n2b, n,n'-ethane-1,2-diyl bis n-acetylacetamide, n-acetyl-n-2-diacetylamino ethyl acetamide | |
BGRWYDHXPHLNKA-UHFFFAOYSA-N | |
N-acetyl-N-[2-(diacetylamino)ethyl]acetamide | |
66347 | |
90% | |
Tetraacetylethylenediamine, 90% |
149°C to 154°C | |
140°C (2.0 hPa) | |
Authentic | |
C10H16N2O4 | |
15, 9158 | |
Solubility in water: ca 1g/L (20°C). Other solubilities: slightly soluble in acetone,moderately soluble in chloroform,methylenechloride | |
CC(=O)N(CCN(C(=O)C)C(=O)C)C(=O)C | |
228.25 | |
228.25 | |
Granular Powder |
Safety and Handling
missing translation for 'einecsNumber' : 234-123-8
missing translation for 'tsca' : TSCA
RUO – Research Use Only